Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTGCAACTTTTATTTTTATCAG[C/T]AGTTGATCCTTAAAGCATCCAAAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600225 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCH1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GCH1 - GTP cyclohydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000161.2 | Intron | NP_000152.1 | ||||
NM_001024024.1 | Intron | NP_001019195.1 | ||||
NM_001024070.1 | Intron | NP_001019241.1 | ||||
NM_001024071.1 | Intron | NP_001019242.1 | ||||
XM_017021218.1 | Intron | XP_016876707.1 |
Set Membership: |
HapMap |