Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACAGTCTATAAAAACTACATAA[A/C]CATGGGGAAAAGCTTACAACAGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602984 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MED6 - mediator complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284209.1 | 1188 | UTR 3 | NP_001271138.1 | |||
NM_001284210.1 | 1188 | UTR 3 | NP_001271139.1 | |||
NM_001284211.1 | 1188 | UTR 3 | NP_001271140.1 | |||
NM_005466.3 | 1188 | UTR 3 | NP_005457.2 |
Set Membership: |
HapMap |