Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGTCTATTCATTACGATCTTT[A/T]TGGAAACTCCTTTATATGGAAACTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603502 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFRD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IFRD1 - interferon related developmental regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007245.2 | Intron | NP_001007246.1 | ||||
NM_001197079.1 | Intron | NP_001184008.1 | ||||
NM_001197080.1 | Intron | NP_001184009.1 | ||||
NM_001550.3 | Intron | NP_001541.2 | ||||
XM_011516142.2 | Intron | XP_011514444.1 | ||||
XM_011516143.2 | Intron | XP_011514445.1 | ||||
XM_017012151.1 | Intron | XP_016867640.1 |