Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATAGGGAGTCATCAACATTGTAG[C/T]TGGAGTAATGGAAATAGAGGAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113810 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEND6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BEND6 - BEN domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318539.1 | Intron | NP_001305468.1 | ||||
NM_152731.2 | Intron | NP_689944.2 | ||||
XM_005248889.1 | Intron | XP_005248946.1 | ||||
XM_011514346.2 | Intron | XP_011512648.1 | ||||
XM_017010406.1 | Intron | XP_016865895.1 | ||||
XM_017010407.1 | Intron | XP_016865896.1 | ||||
XM_017010408.1 | Intron | XP_016865897.1 |
DST - dystonin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |