Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTAAGCGTTCAAAATCCAGTTA[C/T]GTGCTTTGTTATTGAAAACAGTCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153455 MIM: 610479 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOX PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOX - lysyl oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178102.2 | 751 | UTR 3 | NP_001171573.1 | |||
NM_001317073.1 | 751 | UTR 3 | NP_001304002.1 | |||
NM_002317.6 | 751 | UTR 3 | NP_002308.2 |
SRFBP1 - serum response factor binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152546.2 | 751 | Intron | NP_689759.2 | |||
XM_017009111.1 | 751 | Intron | XP_016864600.1 |
Set Membership: |
HapMap |