Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCAGACAAATTGCTCTCTATAA[C/T]GATTTGGCAGATCAATGGAGACATC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125265 MIM: 182175 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
REEP5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
REEP5 - receptor accessory protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005669.4 | 2049 | UTR 3 | NP_005660.4 | |||
XM_017009844.1 | 2049 | Intron | XP_016865333.1 |
SRP19 - signal recognition particle 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204193.1 | 2049 | Intron | NP_001191122.1 | |||
NM_001204194.1 | 2049 | Intron | NP_001191123.1 | |||
NM_001204196.1 | 2049 | Intron | NP_001191125.1 | |||
NM_001204199.1 | 2049 | Intron | NP_001191128.1 | |||
NM_003135.2 | 2049 | Intron | NP_003126.1 |
Set Membership: |
HapMap |