Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAAACATAGAAAGATCTACCCA[C/T]GCATCATATGACTAACATAAAAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607062 MIM: 607774 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FKBP7 - FK506 binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135212.1 | Intron | NP_001128684.1 | ||||
NM_181342.2 | Intron | NP_851939.1 | ||||
XM_005246638.4 | Intron | XP_005246695.1 | ||||
XM_011511348.2 | Intron | XP_011509650.1 | ||||
XM_011511349.2 | Intron | XP_011509651.1 |
PLEKHA3 - pleckstrin homology domain containing A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |