Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCCACTATATCCATCACAGTAT[A/G]GTTGAATGTTACTAGGCCTTGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300535 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OCRL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OCRL - OCRL, inositol polyphosphate-5-phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000276.3 | Intron | NP_000267.2 | ||||
NM_001318784.1 | Intron | NP_001305713.1 | ||||
NM_001587.3 | Intron | NP_001578.2 | ||||
XM_005262422.2 | Intron | XP_005262479.1 | ||||
XM_011531344.2 | Intron | XP_011529646.1 | ||||
XM_011531345.2 | Intron | XP_011529647.1 | ||||
XM_017029554.1 | Intron | XP_016885043.1 |
Set Membership: |
HapMap |