Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTGACTGGAGAGCGGCCGAATT[A/G]GGCACATTTGCGGGAACGCAGAGCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601437 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCGRT PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FCGRT - Fc fragment of IgG receptor and transporter | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCN3 - reticulocalbin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020650.2 | 421 | UTR 5 | NP_065701.2 | |||
XM_005259089.3 | 421 | UTR 5 | XP_005259146.1 | |||
XM_011527143.1 | 421 | UTR 5 | XP_011525445.1 | |||
XM_017027023.1 | 421 | UTR 5 | XP_016882512.1 |
Set Membership: |
HapMap |