Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATGTGCACCCAAAACCAAGGAT[C/T]CATGTGCTCCCCAGGTCAAGAAGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
51 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616363 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRR4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPRR4 - small proline rich protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173080.2 | 891 | Missense Mutation | CCA,TCA | P,S 45 | NP_775103.1 | |
XM_017000482.1 | 891 | Missense Mutation | CCA,TCA | P,S 45 | XP_016855971.1 |