Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTGTTGAGCTATGCTAAGAATT[G/T]ACACTCAGTGGTTGGGCGCGGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606698 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOX30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SOX30 - SRY-box 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308165.1 | Intron | NP_001295094.1 | ||||
NM_007017.2 | Intron | NP_008948.1 | ||||
NM_178424.1 | Intron | NP_848511.1 | ||||
XM_005265803.4 | Intron | XP_005265860.1 | ||||
XM_011534420.1 | Intron | XP_011532722.1 |
Set Membership: |
HapMap |