Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCAGTTGACTCCTAGAAGCAAT[T/G]TGGTCCCTCTGTTCTAAACTGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131220 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FGF1 - fibroblast growth factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000800.4 | 597 | UTR 3 | NP_000791.1 | |||
NM_001144892.2 | 597 | UTR 3 | NP_001138364.1 | |||
NM_001144934.1 | 597 | UTR 3 | NP_001138406.1 | |||
NM_001144935.1 | 597 | UTR 3 | NP_001138407.1 | |||
NM_001257205.1 | 597 | UTR 3 | NP_001244134.1 | |||
NM_001257206.1 | 597 | UTR 3 | NP_001244135.1 | |||
NM_001257207.1 | 597 | UTR 3 | NP_001244136.1 | |||
NM_001257208.1 | 597 | UTR 3 | NP_001244137.1 | |||
NM_001257209.1 | 597 | UTR 3 | NP_001244138.1 | |||
NM_001257210.1 | 597 | UTR 3 | NP_001244139.1 | |||
NM_001257211.1 | 597 | UTR 3 | NP_001244140.1 | |||
NM_001257212.1 | 597 | UTR 3 | NP_001244141.1 | |||
NM_033136.3 | 597 | UTR 3 | NP_149127.1 | |||
NM_033137.2 | 597 | UTR 3 | NP_149128.1 | |||
XM_005268390.4 | 597 | UTR 3 | XP_005268447.1 | |||
XM_011537607.2 | 597 | UTR 3 | XP_011535909.1 |
Set Membership: |
HapMap |