Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTAGGTTAACTTCTAATAAGCAG[A/G]CGAACATGTTACATAAATTATAATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603562 MIM: 609927 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTMR7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MTMR7 - myotubularin related protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004686.4 | 3913 | UTR 3 | NP_004677.3 | |||
XM_006716414.3 | 3913 | Intron | XP_006716477.2 | |||
XM_011544692.2 | 3913 | Intron | XP_011542994.1 | |||
XM_011544693.2 | 3913 | Intron | XP_011542995.1 | |||
XM_017013956.1 | 3913 | UTR 3 | XP_016869445.1 |
VPS37A - VPS37A, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145152.1 | 3913 | UTR 3 | NP_001138624.1 | |||
NM_152415.2 | 3913 | UTR 3 | NP_689628.2 | |||
XM_005273400.2 | 3913 | UTR 3 | XP_005273457.1 | |||
XM_005273401.2 | 3913 | UTR 3 | XP_005273458.1 | |||
XM_006716286.1 | 3913 | UTR 3 | XP_006716349.1 | |||
XM_017013021.1 | 3913 | Intron | XP_016868510.1 | |||
XM_017013022.1 | 3913 | Intron | XP_016868511.1 | |||
XM_017013023.1 | 3913 | UTR 3 | XP_016868512.1 | |||
XM_017013024.1 | 3913 | UTR 3 | XP_016868513.1 | |||
XM_017013025.1 | 3913 | Intron | XP_016868514.1 | |||
XM_017013026.1 | 3913 | UTR 3 | XP_016868515.1 |
Set Membership: |
HapMap |