Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAACCCTGCAATCTGTGACACC[A/G]CACAGGAGCCAGTGGGTTTTGATGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610541 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R3B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPP1R3B - protein phosphatase 1 regulatory subunit 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201329.1 | 1718 | UTR 3 | NP_001188258.1 | |||
NM_024607.3 | 1718 | UTR 3 | NP_078883.2 | |||
XM_006716253.3 | 1718 | UTR 3 | XP_006716316.1 | |||
XM_017013835.1 | 1718 | UTR 3 | XP_016869324.1 |
Set Membership: |
HapMap |