Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGCCAGGAGACCATCCGCGTC[A/C]CCAAGCCCTGCACCCCCAAGACCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118495 MIM: 601441 MIM: 162096 | ||||||||||||||||||||
Literature Links: |
CHRM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRM4 - cholinergic receptor muscarinic 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DGKZ - diacylglycerol kinase zeta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MDK - midkine (neurite growth-promoting factor 2) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012333.2 | 755 | Missense Mutation | ACC,CCC | T,P 123 | NP_001012333.1 | |
NM_001012334.2 | 755 | Missense Mutation | ACC,CCC | T,P 123 | NP_001012334.1 | |
NM_001270550.1 | 755 | Missense Mutation | ACC,CCC | T,P 123 | NP_001257479.1 | |
NM_001270551.1 | 755 | Missense Mutation | ACC,CCC | T,P 123 | NP_001257480.1 | |
NM_001270552.1 | 755 | Missense Mutation | ACC,CCC | T,P 67 | NP_001257481.1 | |
NM_002391.4 | 755 | Missense Mutation | ACC,CCC | T,P 123 | NP_002382.1 | |
XM_011520116.2 | 755 | Missense Mutation | ACC,CCC | T,P 123 | XP_011518418.1 | |
XM_017017764.1 | 755 | Missense Mutation | ACC,CCC | T,P 123 | XP_016873253.1 |
MIR4688 - microRNA 4688 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |