Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGAAACCCAAATGGGCACACT[A/G]AGTCGAAGTCTGAGTCGTCCTTCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607400 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCOLN3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MCOLN3 - mucolipin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253693.1 | Intron | NP_001240622.1 | ||||
NM_018298.10 | Intron | NP_060768.8 | ||||
XM_005271003.1 | Intron | XP_005271060.1 | ||||
XM_006710750.1 | Intron | XP_006710813.1 | ||||
XM_011541739.2 | Intron | XP_011540041.2 | ||||
XM_011541740.2 | Intron | XP_011540042.1 | ||||
XM_011541741.2 | Intron | XP_011540043.1 | ||||
XM_011541742.1 | Intron | XP_011540044.1 |
Set Membership: |
HapMap |