Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGGCTGGGCGCTCCGGCTCCAA[G/T]AGCCTTCGTGTAGAAAAGCTCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605064 MIM: 180520 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF23 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF23 - kinesin family member 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPLP1 - ribosomal protein lateral stalk subunit P1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003.2 | Intron | NP_000994.1 | ||||
NM_213725.1 | Intron | NP_998890.1 |
Set Membership: |
HapMap |