Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAACTATAGTACATCAAAACGTG[A/G]TATTATGCTGCTATGCCATTTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605064 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF23 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF23 - kinesin family member 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281301.1 | Intron | NP_001268230.1 | ||||
NM_004856.6 | Intron | NP_004847.2 | ||||
NM_138555.3 | Intron | NP_612565.1 | ||||
XM_005254796.2 | Intron | XP_005254853.1 | ||||
XM_005254797.2 | Intron | XP_005254854.1 | ||||
XM_005254798.2 | Intron | XP_005254855.1 | ||||
XM_005254799.2 | Intron | XP_005254856.1 | ||||
XM_011522238.1 | Intron | XP_011520540.1 | ||||
XM_011522239.2 | Intron | XP_011520541.1 | ||||
XM_011522240.1 | Intron | XP_011520542.1 | ||||
XM_017022733.1 | Intron | XP_016878222.1 |
LOC145694 - uncharacterized LOC145694 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |