Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGTATGCCGTAGATCTCCATTGT[C/G]TAGGCCTAGTTAATCATCGCCGGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614476 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CT62 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CT62 - cancer/testis antigen 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102658.1 | 1406 | UTR 3 | NP_001096128.1 | |||
XM_006720429.3 | 1406 | UTR 3 | XP_006720492.1 |
THSD4 - thrombospondin type 1 domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |