Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGCAGCTATTTCAAATGTTCCA[A/G]ATTTCTCTAAACTGCTACTATTTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602331 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ORC5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ORC5 - origin recognition complex subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002553.3 | Intron | NP_002544.1 | ||||
NM_181747.3 | Intron | NP_859531.1 | ||||
XM_011516273.2 | Intron | XP_011514575.1 |
Set Membership: |
HapMap |