Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGCCTAACTGATAGAATCTTTC[C/T]AAGCTGGTTTATACTAGTTCCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D31 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TBC1D31 - TBC1 domain family member 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145088.1 | Intron | NP_001138560.1 | ||||
NM_145647.3 | Intron | NP_663622.2 | ||||
XM_005251102.1 | Intron | XP_005251159.1 | ||||
XM_005251103.1 | Intron | XP_005251160.1 | ||||
XM_005251104.1 | Intron | XP_005251161.1 | ||||
XM_011517378.2 | Intron | XP_011515680.1 | ||||
XM_011517379.2 | Intron | XP_011515681.1 | ||||
XM_017013983.1 | Intron | XP_016869472.1 | ||||
XM_017013984.1 | Intron | XP_016869473.1 | ||||
XM_017013985.1 | Intron | XP_016869474.1 | ||||
XM_017013986.1 | Intron | XP_016869475.1 | ||||
XM_017013987.1 | Intron | XP_016869476.1 | ||||
XM_017013988.1 | Intron | XP_016869477.1 | ||||
XM_017013989.1 | Intron | XP_016869478.1 | ||||
XM_017013990.1 | Intron | XP_016869479.1 |