Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGAAATCTAACTTATTTTCTTAC[A/G]CTAAAACATTGGCAATAAGAGATTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610617 MIM: 611350 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTL PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DTL - denticleless E3 ubiquitin protein ligase homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286229.1 | Intron | NP_001273158.1 | ||||
NM_001286230.1 | Intron | NP_001273159.1 | ||||
NM_016448.3 | Intron | NP_057532.3 | ||||
XM_011509614.1 | Intron | XP_011507916.1 |
INTS7 - integrator complex subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |