Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGTGAAGGTGTTCCGGGCGGC[C/T]GACCCGCTGGTGGGTGTGTTCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602821 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF5A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF5A - kinesin family member 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIP4K2C - phosphatidylinositol-5-phosphate 4-kinase type 2 gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146258.1 | 256 | Silent Mutation | GCC,GCT | A,A 44 | NP_001139730.1 | |
NM_001146259.1 | 256 | Silent Mutation | GCC,GCT | A,A 44 | NP_001139731.1 | |
NM_001146260.1 | 256 | Silent Mutation | GCC,GCT | A,A 44 | NP_001139732.1 | |
NM_024779.4 | 256 | Silent Mutation | GCC,GCT | A,A 44 | NP_079055.3 | |
XM_005269152.2 | 256 | Silent Mutation | GCC,GCT | A,A 44 | XP_005269209.1 | |
XM_011538747.1 | 256 | Silent Mutation | GCC,GCT | A,A 44 | XP_011537049.1 | |
XM_017019973.1 | 256 | UTR 5 | XP_016875462.1 |
Set Membership: |
HapMap |