Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGGAAAGCGAGCCTGGAGGGC[A/G]CAGACGGACGACTAGCTGTGCTCGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604482 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NME2P1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NME2P1 - NME/NM23 nucleoside diphosphate kinase 2 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNU4-1 - RNA, U4 small nuclear 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNU4-2 - RNA, U4 small nuclear 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRT4 - sirtuin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012240.2 | 19 | Intron | NP_036372.1 | |||
XM_005253865.4 | 19 | Intron | XP_005253922.1 | |||
XM_006719308.2 | 19 | UTR 5 | XP_006719371.1 | |||
XM_006719309.3 | 19 | Intron | XP_006719372.1 |