Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCCACTGTTAATGCCACATACC[A/T]GTGAATGCAAAGTCACTGCAGTTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603484 MIM: 608552 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRC1 - protein regulator of cytokinesis 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS33B - VPS33B, late endosome and lysosome associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289148.1 | 1947 | Silent Mutation | NP_001276077.1 | |||
NM_001289149.1 | 1947 | Silent Mutation | NP_001276078.1 | |||
NM_018668.4 | 1947 | Silent Mutation | NP_061138.3 | |||
XM_005254887.1 | 1947 | Silent Mutation | XP_005254944.1 | |||
XM_011521448.1 | 1947 | Silent Mutation | XP_011519750.1 | |||
XM_011521449.2 | 1947 | Silent Mutation | XP_011519751.1 | |||
XM_017022075.1 | 1947 | Silent Mutation | XP_016877564.1 | |||
XM_017022076.1 | 1947 | Silent Mutation | XP_016877565.1 |
Set Membership: |
HapMap |