Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTTGATTTACATTTCTCTTGAG[A/G]TCTGAATTCACACAGAGCTGCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600594 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGCR11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DGCR11 - DiGeorge syndrome critical region gene 11 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DGCR2 - DiGeorge syndrome critical region gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173533.1 | Intron | NP_001167004.1 | ||||
NM_001173534.1 | Intron | NP_001167005.1 | ||||
NM_001184781.1 | Intron | NP_001171710.1 | ||||
NM_005137.2 | Intron | NP_005128.1 |