Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGTGGATGGTTTCATTCAATC[A/C]ATGTTCGACCAGCTTTATATCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603086 MIM: 607607 | ||||||||||||||||||||
Literature Links: |
ART3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ART3 - ADP-ribosyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP54 - nucleoporin 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278603.1 | 1598 | Nonsense Mutation | GGA,TGA | G,* 446 | NP_001265532.1 | |
NM_017426.3 | 1598 | Nonsense Mutation | GGA,TGA | G,* 494 | NP_059122.2 | |
XM_006714237.1 | 1598 | Nonsense Mutation | GGA,TGA | G,* 314 | XP_006714300.1 | |
XM_011532033.2 | 1598 | Nonsense Mutation | GGA,TGA | G,* 458 | XP_011530335.1 | |
XM_011532034.2 | 1598 | Nonsense Mutation | GGA,TGA | G,* 314 | XP_011530336.1 | |
XM_017008294.1 | 1598 | Nonsense Mutation | GGA,TGA | G,* 314 | XP_016863783.1 | |
XM_017008295.1 | 1598 | Nonsense Mutation | GGA,TGA | G,* 314 | XP_016863784.1 |