Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTCTTACCTACTAGAATATAT[C/T]TTCCTTTCTATTTACTGGGAGGCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601591 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKG2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRKG2 - protein kinase, cGMP-dependent, type II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282480.1 | 2201 | UTR 3 | NP_001269409.1 | |||
NM_001282481.1 | 2201 | UTR 3 | NP_001269410.1 | |||
NM_001282482.1 | 2201 | UTR 3 | NP_001269411.1 | |||
NM_001282483.1 | 2201 | UTR 3 | NP_001269412.1 | |||
NM_001282485.1 | 2201 | UTR 3 | NP_001269414.1 | |||
NM_006259.2 | 2201 | UTR 3 | NP_006250.1 | |||
XM_005263126.3 | 2201 | UTR 3 | XP_005263183.1 | |||
XM_017008413.1 | 2201 | Intron | XP_016863902.1 | |||
XM_017008414.1 | 2201 | Intron | XP_016863903.1 | |||
XM_017008415.1 | 2201 | Intron | XP_016863904.1 | |||
XM_017008416.1 | 2201 | Intron | XP_016863905.1 | |||
XM_017008417.1 | 2201 | Intron | XP_016863906.1 | |||
XM_017008418.1 | 2201 | Intron | XP_016863907.1 |
Set Membership: |
HapMap |