Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAACTATCCACATAGACTTAGA[C/T]AGTAACTTTAGGCTATTCAATCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613568 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMYM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZMYM4 - zinc finger MYM-type containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005095.2 | Intron | NP_005086.2 | ||||
XM_005271328.4 | Intron | XP_005271385.1 | ||||
XM_005271331.2 | Intron | XP_005271388.1 | ||||
XM_011542421.2 | Intron | XP_011540723.1 | ||||
XM_011542423.2 | Intron | XP_011540725.1 | ||||
XM_011542425.2 | Intron | XP_011540727.1 | ||||
XM_017002803.1 | Intron | XP_016858292.1 | ||||
XM_017002804.1 | Intron | XP_016858293.1 | ||||
XM_017002805.1 | Intron | XP_016858294.1 |
Set Membership: |
HapMap |