Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCATGGGTTTTGGAGGGCCTCC[A/G]TTTACAAAGGATGAATTACAATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMPDL3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SMPDL3B - sphingomyelin phosphodiesterase acid like 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009568.2 | Intron | NP_001009568.1 | ||||
NM_001304579.1 | Intron | NP_001291508.1 | ||||
NM_014474.3 | Intron | NP_055289.2 | ||||
XM_011541259.2 | Intron | XP_011539561.1 | ||||
XM_011541260.2 | Intron | XP_011539562.1 |
Set Membership: |
HapMap |