Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATAACATGTTGAACTTAACCAA[A/G]CCTGCTTGCAAAAGGGTAGACATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609101 MIM: 608048 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXO30 - F-box protein 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032145.4 | 6067 | Intron | NP_115521.3 | |||
XM_005267159.3 | 6067 | Intron | XP_005267216.1 | |||
XM_011536177.2 | 6067 | Intron | XP_011534479.1 | |||
XM_017011353.1 | 6067 | Intron | XP_016866842.1 |
LOC100507557 - uncharacterized LOC100507557 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHPRH - SNF2 histone linker PHD RING helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042683.2 | 6067 | Intron | NP_001036148.2 | |||
NM_173082.3 | 6067 | Intron | NP_775105.1 | |||
XM_006715439.3 | 6067 | UTR 3 | XP_006715502.1 | |||
XM_006715441.3 | 6067 | Intron | XP_006715504.1 | |||
XM_006715443.3 | 6067 | Intron | XP_006715506.1 | |||
XM_011535719.2 | 6067 | Intron | XP_011534021.1 | |||
XM_011535722.2 | 6067 | Intron | XP_011534024.1 | |||
XM_017010691.1 | 6067 | Intron | XP_016866180.1 | |||
XM_017010692.1 | 6067 | Intron | XP_016866181.1 | |||
XM_017010693.1 | 6067 | Intron | XP_016866182.1 | |||
XM_017010694.1 | 6067 | Intron | XP_016866183.1 | |||
XM_017010695.1 | 6067 | Intron | XP_016866184.1 | |||
XM_017010696.1 | 6067 | Intron | XP_016866185.1 |
Set Membership: |
HapMap |