Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAAAATCAACAAACATTTCTTTC[C/T]GTGGTCCTTTGGAGGAATGTAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCN3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCN3A - sodium voltage-gated channel alpha subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081676.1 | Intron | NP_001075145.1 | ||||
NM_001081677.1 | Intron | NP_001075146.1 | ||||
NM_006922.3 | Intron | NP_008853.3 | ||||
XM_011511610.2 | Intron | XP_011509912.1 | ||||
XM_011511613.2 | Intron | XP_011509915.1 | ||||
XM_017004660.1 | Intron | XP_016860149.1 | ||||
XM_017004661.1 | Intron | XP_016860150.1 | ||||
XM_017004662.1 | Intron | XP_016860151.1 | ||||
XM_017004663.1 | Intron | XP_016860152.1 | ||||
XM_017004664.1 | Intron | XP_016860153.1 | ||||
XM_017004665.1 | Intron | XP_016860154.1 | ||||
XM_017004666.1 | Intron | XP_016860155.1 |