Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAAAAAAAAAACACGGTTCCT[A/G]TTTTTCAAAGACACTGGACTCCCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604127 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBX15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TBX15 - T-box 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152380.2 | 2358 | UTR 3 | NP_689593.2 | |||
XM_005271161.3 | 2358 | UTR 3 | XP_005271218.1 | |||
XM_005271162.1 | 2358 | UTR 3 | XP_005271219.1 | |||
XM_011542052.2 | 2358 | UTR 3 | XP_011540354.1 |
Set Membership: |
HapMap |