Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGACAGTCACTGGCTCTTTCA[C/T]GAGGAGCCTGGAGAGGGCAGAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607141 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLIPR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLIPR2 - GLI pathogenesis related 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287010.1 | Intron | NP_001273939.1 | ||||
NM_001287011.1 | Intron | NP_001273940.1 | ||||
NM_001287012.1 | Intron | NP_001273941.1 | ||||
NM_001287013.1 | Intron | NP_001273942.1 | ||||
NM_001287014.1 | Intron | NP_001273943.1 | ||||
NM_022343.3 | Intron | NP_071738.1 |
Set Membership: |
HapMap |