Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCTGCACACCACTTTAGTCTT[C/T]AGCCATATCAGCTGCTGATATGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611854 MIM: 603132 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL50 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MRPL50 - mitochondrial ribosomal protein L50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF189 - zinc finger protein 189 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278231.1 | Intron | NP_001265160.1 | ||||
NM_001278232.1 | Intron | NP_001265161.1 | ||||
NM_001278240.1 | Intron | NP_001265169.1 | ||||
NM_003452.3 | Intron | NP_003443.2 | ||||
NM_197977.2 | Intron | NP_932094.1 | ||||
XM_006717280.3 | Intron | XP_006717343.1 | ||||
XM_006717281.3 | Intron | XP_006717344.1 | ||||
XM_011518998.2 | Intron | XP_011517300.1 | ||||
XM_011518999.2 | Intron | XP_011517301.1 | ||||
XM_017015121.1 | Intron | XP_016870610.1 |
Set Membership: |
HapMap |