Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGATGCTATATATTAACTAACA[C/T]AGACATATCACCTTTCATTTTGTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601434 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SKP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
AMR
|
||||||||
EUR - Not Available |
SKP1 - S-phase kinase-associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006930.3 | Intron | NP_008861.2 | ||||
NM_170679.2 | Intron | NP_733779.1 |