Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTCTGGGTCAAAAATTTTGTT[A/G]AGAGCTTTGCTAAGCACTTAACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MMP27 - matrix metallopeptidase 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022122.2 | 2217 | Intron | NP_071405.2 | |||
XM_011542948.2 | 2217 | UTR 3 | XP_011541250.1 | |||
XM_011542949.2 | 2217 | UTR 3 | XP_011541251.1 | |||
XM_011542950.2 | 2217 | UTR 3 | XP_011541252.1 | |||
XM_017018120.1 | 2217 | UTR 3 | XP_016873609.1 |
Set Membership: |
HapMap |