Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGCTGTTTTAAAATGGCAGAAA[C/T]GATGGAATGTTTACTCTGTCTAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612297 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1549L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIAA1549L - KIAA1549 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012194.2 | Intron | NP_036326.2 | ||||
XM_005252847.3 | Intron | XP_005252904.2 | ||||
XM_005252848.3 | Intron | XP_005252905.1 | ||||
XM_011519969.2 | Intron | XP_011518271.1 | ||||
XM_011519970.2 | Intron | XP_011518272.1 | ||||
XM_017017484.1 | Intron | XP_016872973.1 | ||||
XM_017017485.1 | Intron | XP_016872974.1 | ||||
XM_017017486.1 | Intron | XP_016872975.1 |