Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTGTCTTCTGTTTTCAGATTAA[C/G]GGTTGAAAATTTCCCATTAAAATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603341 MIM: 600024 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAH14 - dynein axonemal heavy chain 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LBR - lamin B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002296.3 | 2107 | UTR 3 | NP_002287.2 | |||
NM_194442.2 | 2107 | UTR 3 | NP_919424.1 | |||
XM_005273125.2 | 2107 | UTR 3 | XP_005273182.1 | |||
XM_011544185.2 | 2107 | UTR 3 | XP_011542487.1 |
Set Membership: |
HapMap |