Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTTGGGGTTACCAGACACATTC[C/T]ATCTCCTTCTCTCTCTCCATCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602188 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EPHA4 - EPH receptor A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304536.1 | 4886 | UTR 3 | NP_001291465.1 | |||
NM_001304537.1 | 4886 | UTR 3 | NP_001291466.1 | |||
NM_004438.4 | 4886 | UTR 3 | NP_004429.1 | |||
XM_005246374.2 | 4886 | UTR 3 | XP_005246431.1 |
Set Membership: |
HapMap |