Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTTCCCTCCAGGAACTGATCCT[C/T]GGGAAAACAGCCATGGGCCAGGCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614827 MIM: 612532 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAJC11 - DnaJ heat shock protein family (Hsp40) member C11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018198.3 | 1879 | UTR 3 | NP_060668.2 | |||
XM_017001745.1 | 1879 | UTR 3 | XP_016857234.1 |
THAP3 - THAP domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195752.1 | 1879 | Intron | NP_001182681.1 | |||
NM_001195753.1 | 1879 | Intron | NP_001182682.1 | |||
NM_138350.3 | 1879 | UTR 3 | NP_612359.2 | |||
XM_005263532.3 | 1879 | Intron | XP_005263589.1 | |||
XM_011542400.2 | 1879 | Intron | XP_011540702.1 | |||
XM_011542401.2 | 1879 | Intron | XP_011540703.1 | |||
XM_017002761.1 | 1879 | UTR 3 | XP_016858250.1 |
Set Membership: |
HapMap |