Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCGTCGCGCCGGGTCCCGCTTC[A/C]GCTGCTGCTGCTCGGCGGCCTTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 135820 | ||||||||||||||||||||
Literature Links: |
FBLN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBLN1 - fibulin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001996.3 | 185 | Missense Mutation | CAG,CCG | Q,P 13 | NP_001987.2 | |
NM_006485.3 | 185 | Missense Mutation | CAG,CCG | Q,P 13 | NP_006476.2 | |
NM_006486.2 | 185 | Missense Mutation | CAG,CCG | Q,P 13 | NP_006477.2 | |
NM_006487.2 | 185 | Missense Mutation | CAG,CCG | Q,P 13 | NP_006478.2 |