Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTTTAGTCATTCATTCTAAATA[C/T]TTCTCCATTTTCAACCTGTCAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612186 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMXL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DMXL2 - Dmx like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174116.1 | Intron | NP_001167587.1 | ||||
NM_001174117.1 | Intron | NP_001167588.1 | ||||
NM_015263.3 | Intron | NP_056078.2 | ||||
XM_005254255.1 | Intron | XP_005254312.1 | ||||
XM_005254256.1 | Intron | XP_005254313.1 | ||||
XM_011521402.2 | Intron | XP_011519704.1 | ||||
XM_017022034.1 | Intron | XP_016877523.1 |