Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGCTCCAGGCGGAGGCATCGC[A/C]GCCGAGGTAAACTCAGGGAACTTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602706 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRDE2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NRDE2 - NRDE-2, necessary for RNA interference, domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017970.3 | Intron | NP_060440.2 | ||||
XM_011536886.1 | Intron | XP_011535188.1 | ||||
XM_017021394.1 | Intron | XP_016876883.1 | ||||
XM_017021395.1 | Intron | XP_016876884.1 | ||||
XM_017021396.1 | Intron | XP_016876885.1 |
PSMC1 - proteasome 26S subunit, ATPase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |