Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATGGATGAATTCTGCCTTTCTC[C/T]GTTGGTAGCCATTGTGTTGTGCACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP44 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGAP44 - Rho GTPase activating protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321164.1 | Intron | NP_001308093.1 | ||||
NM_001321166.1 | Intron | NP_001308095.1 | ||||
NM_001321167.1 | Intron | NP_001308096.1 | ||||
NM_001321168.1 | Intron | NP_001308097.1 | ||||
NM_014859.5 | Intron | NP_055674.4 | ||||
XM_005256891.1 | Intron | XP_005256948.1 |
LOC100128006 - uncharacterized LOC100128006 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |