Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCAATTGTATGTACAGAGGCTT[C/G]ATTAGTTTTCCCAAGAACTTCCAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180247 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RXRG PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RXRG - retinoid X receptor gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256570.1 | 1577 | UTR 3 | NP_001243499.1 | |||
NM_001256571.1 | 1577 | UTR 3 | NP_001243500.1 | |||
NM_006917.4 | 1577 | UTR 3 | NP_008848.1 |
Set Membership: |
HapMap |