Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTTTGTTCAAGATCAAATGGA[C/T]GTGATTTGCCTTGTGGAGGTGGCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD46 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD46 - ankyrin repeat domain 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270377.1 | 1207 | Intron | NP_001257306.1 | |||
NM_001270378.1 | 1207 | Intron | NP_001257307.1 | |||
NM_001270379.1 | 1207 | UTR 3 | NP_001257308.1 | |||
NM_198401.3 | 1207 | Intron | NP_940683.1 | |||
XM_006716509.1 | 1207 | Intron | XP_006716572.1 | |||
XM_017013106.1 | 1207 | Intron | XP_016868595.1 |