Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTGGTGATAGTTGTCATGGGAA[C/T]GGAAGTCAGGGGCAATGGATTCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608384 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSDMC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GSDMC - gasdermin C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031415.2 | Intron | NP_113603.1 | ||||
XM_011517158.1 | Intron | XP_011515460.1 | ||||
XM_011517159.1 | Intron | XP_011515461.1 | ||||
XM_011517161.1 | Intron | XP_011515463.1 | ||||
XM_011517162.1 | Intron | XP_011515464.1 | ||||
XM_011517163.2 | Intron | XP_011515465.1 | ||||
XM_011517165.1 | Intron | XP_011515467.1 | ||||
XM_011517166.2 | Intron | XP_011515468.1 | ||||
XM_011517167.1 | Intron | XP_011515469.1 | ||||
XM_011517168.1 | Intron | XP_011515470.1 | ||||
XM_017013644.1 | Intron | XP_016869133.1 | ||||
XM_017013645.1 | Intron | XP_016869134.1 |