Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCTTAGGATCAAAGAAGACTCA[G/T]TGGTGTATAGAGTAAGCCCTGAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600124 MIM: 604135 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPA2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HNRNPA2B1 - heterogeneous nuclear ribonucleoprotein A2/B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002137.3 | 3079 | UTR 3 | NP_002128.1 | |||
NM_031243.2 | 3079 | UTR 3 | NP_112533.1 | |||
XM_005249729.1 | 3079 | Intron | XP_005249786.1 | |||
XM_017012109.1 | 3079 | UTR 3 | XP_016867598.1 | |||
XM_017012110.1 | 3079 | Intron | XP_016867599.1 |
NFE2L3 - nuclear factor, erythroid 2 like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |