Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGTCTACATTTACCATACCTTA[G/T]CCATGAGTGTTGTTCCAAATCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608475 | ||||||||||||||||||||
Literature Links: |
HIBADH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIBADH - 3-hydroxyisobutyrate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152740.3 | 1091 | Missense Mutation | GAT,GCT | D,A 283 | NP_689953.1 | |
XM_017011719.1 | 1091 | Missense Mutation | GAT,GCT | D,A 182 | XP_016867208.1 |